About this item
This MAb recognizes the 56.5 kDa (CK10); 50 kDa (CK14); 50 kDa (CK15); 48 kDa (CK16); 40 kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Type: Primary
Antigen: KRT77
Clonality: Monoclonal
Clone: Clone KRTL/1077
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75915-970
- 75915-972
- Antigen name:
- Cytokeratin LMW
- Cytokeratin LMW
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- KRTL/1077
- KRTL/1077
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 654392
- Unigene ID: 654392
- Isotype:
- IgG1 kappa
- IgG1 kappa