About this item
The antibody HEB-29 reacts with human blood group B. The specificity of the antibody HEB-29 was confirmed by comparison of specificity and reactivity to standard reagent using >5.000 samples of blood. MAb HEB-29 shows specific staining of erythrocytes and vascular epithelium of blood group B controls and no staining in group A controls. It is applicable for tissue staining in tumor patients with blood groups B and AB. Blood group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: HEB-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75885-412
- 75885-414
- Antigen name:
- B
- B
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HEB-29
- HEB-29
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 654423
- Unigene ID: 654423
- Isotype:
- IgM kappa
- IgM kappa