About this item
Cytochrome c is a well-characterized mobile electron transport protein that is essential to energy conversion in all aerobic organisms. In mammalian cells, this highly conserved protein is normally localized to the mitochondrial inter-membrane space. More recent studies have identified cytosolic Cytochrome c as a factor necessary for activation of apoptosis. During apoptosis, Cytochrome c is trans-located from the mitochondrial membrane to the cytosol, where it is required for activation of caspase-3 (CPP32). Overexpression of Bcl-2 has been shown to prevent the translocation of Cytochrome c, thereby blocking the apoptotic process. Overexpression of Bax has been shown to induce the release of Cytochrome c and to induce cell death. The release of Cytochrome c from the mitochondria is thought to trigger an apoptotic cascade, whereby Apaf-1 binds to Apaf-3 (caspase-9) in a Cytochrome c-dependent manner, leading to caspase-9 cleavage of caspase-3. This MAb recognizes total Cytochrome c which includes both apocytochrome (i.e. cytochrome in the cytosol without heme attached) and holocytochrome (i.e cytochrome in the mitochondria with heme attached).
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: Clone CYCS/1010
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75923-920
- 75923-922
- Antigen name:
- Cytochrome C
- Cytochrome C
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- CYCS/1010
- CYCS/1010
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 437060
- Unigene ID: 437060
- Isotype:
- IgG2b kappa
- IgG2b kappa