About this item
This antibody recognizes the CD45 leukocyte common antigen (LCA) family which comprises at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant non-hematopoietic tissues. The intracellular portions of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals. Antibody to CD45 is useful in differential diagnosis of lymphoid tumors from non-hematopoietic undifferentiated neoplasms. A positive result with this MAb is highly indicative of lymphoid or myeloid origin. Certain types of lymphoid neoplasms may lack CD45 (Hodgkin lymphoma, some T-cell lymphomas, and some leukemias) so its absence does not rule out a hematolymphoid tumor. This antibody is expressed almost exclusively by cells of hematopoietic lineage and is present in most benign and malignant lymphocytes as well as plasma cell precursors.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: Clone 2B11 PD7/26
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Dog, Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75912-888
- 75912-886
- Antigen name:
- CD45 / LCA
- CD45 / LCA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 2B11 PD7/26
- 2B11 PD7/26
- Host:
- Mouse
- Mouse
- Gene ID:
- Unigene ID: 654514
- Unigene ID: 654514
- Isotype:
- IgG
- IgG