Order Entry
ContactUsLinkComponent
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 109-3C2]
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 109-3C2]
Catalog #: 75890-534
Supplier:  Biotium
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 109-3C2]
Catalog #: 75890-534
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD20
  • Clonality:
    Monoclonal
  • Clone:
    109-3C2
  • Gene ID:
    Unigene ID: 712553
  • Host:
    Mouse
  • Isotype:
    IgG3 kappa
  • Reactivity:
    Human
  • Antigen symbol:
    MS4A1
  • Conjugation:
    Unconjugated
  • ImmunoFluorescence:
    Yes
  • Size:
    50 μL
  • Storage buffer:
    PBS, no BSA, no azide
  • Concentration:
    1 mg/mL
  • Cat. no.:
    75890-534
  • Supplier No.:
    BNUM0355-50

Specifications

About this item

This antibody recognizes a protein of 30-33 kDa, which is identified as CD20 (Workshop V; Code CD20.12. Workshop IV; Code B17). It This antibody recognizes an extracellular domain of CD20. It is a non-Ig differentiation antigen of B-cells and its expression is restricted to normal and neoplastic B-cells, being absent from all other leukocytes and tissues. CD20 is expressed by pre B-cells and persists during all stages of B-cell maturation but is lost upon terminal differentiation into plasma cells. The protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. CD20 is not glycosylated in resting B-cells and its cytoplasmic domains are differentially phosphorylated upon activation. It acts as calcium channel involved in B cell activation and cell cycle progression.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: Clone 109-3C2
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human

Customers Who Bought This Also Bought