About this item
This MAb is highly specific to testosterone. Its affinity constant for testosterone is ~1010 M-1. In competitive binding immunoassay, it reacts with testosterone 100%, 11-beta-hydroxy testosterone 3.3%, 17-alpha-methyl testosterone <0.1%, 5-alpha DHT 0.8%, estradiol <0.1% and progesterone <0.1%. Testosterone, a steroid hormone from the androgen group, is derived from cholesterol. Testosterone is primarily secreted in the testes, but the ovaries and adrenal glands also produce smaller amounts. It is the principal male sex hormone and is responsible for the maturation of sex organs and the development of male secondary sex characteristics. The hormone is also involved in the growth of muscle mass and increases bone density and maturation. It functions by activating the androgen receptor and converting to estradiol and activation of estrogen receptors. Testosterone has been employed therapeutically to treat many conditions including infertility, lack of libido, osteoporosis, height growth and bone marrow stimulation. Anti-testosterone antibody may also prove useful in identification of testicular tumors arising from interstitial cells.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone 4E1G2
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: All species
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 75974-942
- 75974-940
- Antigen name:
- Testosterone
- Testosterone
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 4E1G2
- 4E1G2
- Reactivity:
- Multiple
- Multiple
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa