To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SHBG
- Clonality:Monoclonal
- Clone:SHBG/245
- Gene ID:Unigene ID: 632235
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:SHBG
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μL
- Storage buffer:PBS, no BSA, no azide
- Concentration:1 mg/mL
- Cat. no.:75974-168
- Supplier No.:BNUM0245-50
Specifications
About this item
This antibody recognizes a protein of 45 kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis.
Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: Clone SHBG/245
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human