Order Entry
ContactUsLinkComponent
Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12]
Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12]
Catalog #: 75967-564
Supplier:  Biotium
Anti-TP53 Mouse Monoclonal Antibody [clone: BP53-12]
Catalog #: 75967-564
Supplier:  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor protein p53
  • Clonality:
    Monoclonal
  • Clone:
    BP53-12
  • Gene ID:
    Unigene ID: 654481
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Hamster,
    Monkey,
    Chicken,
    Human,
    Dog
  • Antigen symbol:
    TP53
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    50 μL
  • Western blot:
    Yes
  • Storage buffer:
    PBS, no BSA, no azide
  • Concentration:
    1 mg/mL
  • Cat. no.:
    75967-564
  • Supplier No.:
    BNUM0013-50

Specifications

About this item

This MAb reacts with an N-terminal epitope (aa 16-25) of both wild type and mutated p53. Mutation and/or allelic loss of p53 is one of the causes of a variety of mesenchymal and epithelial tumors. If it occurs in the germ line, such tumors run in families. In most transformed and tumor cells the concentration of p53 is increased 51000 fold over the minute concentrations (1000 molecules cell) in normal cells, principally due to the increased half-life (4 h) compared to that of the wild-type (20 min). p53 Localizes in the nucleus, but is detectable at the plasma membrane during mitosis and when certain mutations modulate cytoplasmic/nuclear distribution. Mutations arise with an average frequency of 70% but incidence varies from zero in carcinoid lung tumors to 97% in primary melanomas. High concentrations of p53 protein are transiently expressed in human epidermis and superficial dermal fibroblasts following mild ultraviolet irradiation. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia.

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: Clone BP53-12
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Dog, Hamster, Human, Monkey

Customers Who Bought This Also Bought