Neulog Sense Servo Gripper Arm
Catalog #: 470232-638
Supplier: Wards
Specifications
- Description:Neulog Sense Servo Gripper Arm
- Cat. no.:470232-638
Specifications
About this item
An accessory for your SENSE bot!
- Gripper turns sense into a robot that can modify its environment
- The gripper has 2 servomotors, one to move the gripper up and down, the other to open and close
- Each servo of the gripper has a transmission gear and potentiometer
- Requires a Neulog SENSE to run
- Gripper received various commands you can program: up-mid-down, open, close
Allow your Sense to monitor and change its environment with the Sense Gripper. The gripper has two servomotors, one to move the gripper up and down, the other to open and close. Each servo of the gripper has a transmission gear and potentiometer. With block programming it is easy to operate the gripper with commands within the block code. An example of these commands are Up-Mid-Down, Open-Close. Be creative and activate the gripper using other Neulog sensors you can plug into sense, for example use the sound sensor to activate the gripper with a clap.
Customers Who Bought This Also Bought