Neulog Sense Infrared Transmitter
Catalog #: 470232-636
Supplier: Wards
Specifications
- Description:Neulog Sense Infrared Transmitter
- Cat. no.:470232-636
Specifications
About this item
An accessory for your SENSE bot!
- Neulog sense infrared transmitting unit
- Great for sending an IR sensor to use in group robotic events, such as building Sense bot traffic, or battle bots
- Easy USB connection to Neulog Sense
- Recommended for use with the Neulog Sense Tracking Unit which can be purchased separately
The Neulog Sense infrared transmitter is the sister to the tracking unit. The transmitter outputs an IR light that the Neulog Sense infrared tracking unit can locate. It is great for coding robots to follow IR signals or build follow and chase scenarios with multiple sense bots. Most commonly used in an autonomous car challenge where one Sense bot needs to follow another, much like adaptive cruise control in many of todays modern cars.
Customers Who Bought This Also Bought