Specifications
- Description:Neulog Sense Make
- Cat. no.:470232-644
Specifications
About this item
A build and program your own robot system.
- Sense make is for the DIYer or advanced student who would like to design their robot, build it, and program it from scratch
The ROBO206 included has three input for connecting switches and sensors, and three outputs for connection of lamps, motors, indicating LEDs, push button switch and USB mini connection for data transmission from the PC to your robot. Neulog sensors can also be plugged directly into ROBO206.
The coding interface can be downloaded free of charge on the Neulog site as indicated in the SENSEMAKE package.
SENSEMAKE connects directly to your PC via ROBO206 and a USB cable. ROBO206 can be used as USB module for Neulog sensors with Neulog software
Sense make is great for advanced students, maker spaces, robot competitions, or the home DIY person that wants to design, build, and code a robot from scratch. The program interface is free for download from the Neulog site as indicated in the Sensemake package. SENSE make has an advanced controller unit to accept inputs and give outputs. Any of the 45 Neulog sensors can be plugged into SENSEMAKE and programmed as desired.