β-(N-Morpholino)ethanesulfonic acid (MES) ≥99% (by titrimetric analysis on dry substance), ULTROL®, Calbiochem®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: 2-(N-Morpholino)ethanesulphonic acid,2-(N-Morpholino)ethanesulfonic acid,2-Morpholinoethanesulfonic acid
Formula:C₆H₁₃NO₄S
MW:195.24 g/mol
MDL Number:MFCD00006181
CAS Number:4432-31-9
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 316 °C
:Flash Pt:
:Flash Pt:
:M.W. 195.24 g/mol
:316
Storage Temperature:Ambient
Customers Who Bought This Also Bought