Specifications
- Description:ToxinEraser™ Endotoxin Resin Removal Kit, Resin Volume of 2 mL of 50% Slurry
- Cat. no.:89233-330
- Supplier No.:L00338
Specifications
About this item
The ToxinEraser™ endotoxin removal resin utilizes immobilized polymixin B to bind and remove pyrogens from solution.
- High stability and removal efficiency
- Excellent binding capacity of over 2000000 EU/mL (CV)
- Strong resilience
The polymixins are a family of antibiotics that contain a cationic cyclopeptide with a fatty acid chain. Polymixin B neutralizes the biological activity of endotoxins by binding to the lipid A portion of bacterial lipopolysaccharide.