PIPETMAN® Diamond Tips EcoPack™, Gilson®
Supplier: GILSON, INC.
Certificates
About this item
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes.
- Accurate and precise: Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results
- Reliable: Our tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your experiments
- Economical: Ordering in bulk and simply reloading the tips into empty racks saves you money
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN perfectly. They deliver accurate and precise pipetting when used in conjunction with Gilson pipettes. ECOPACK™ is an economical way to order tips.
Packaging: Each ECOPACK contains 10,000 fully-autoclavable tips (10 bags of 1000 tips). The bags are sealed with a tamper-proof, rapid-sealing strip for easy access and to prevent contamination and loss of tips. Color-coding on the label enables easy identification and compliance with Good Laboratory Practice (GLP).
Customers Who Bought This Also Bought