About this item
Designed for today's crowded laboratory benchtops.
- Takes up less bench space—40% smaller than comparable models
- Super-large dual-line display is easy to read
- Simple calibrations with 5-point automatic pH buffer recognition for USA and NIST buffers
- More accurate measurements with up to 5-point calibration for ion concentration on Ion 700 model
- Versatile—use with any pH, ORP, or ISE electrodes with BNC connector
Ion 700 meter measures pH, mV, relative mV, and temperature, like a standard benchtop pH meter. When used with ISE's for ion concentration measurements, it also reads directly in ppm for easy use. Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures.
Meters feature Auto-Hold, selectable °F or °C measurement, and easy recall of electrode slope or offset.
Model WD-35419-22 includes meter with universal AC power adapter, and a removeable electrode stand. For replacement parts, see model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.
Model WD-35419-23 includes meter with universal AC power adapter, and one All-in-One pH/ATC electrode. For replacement parts, see model WD-35811-71 for the probe, model WD-35615-07 for the univerrsal power adapter, and model WD-35420-54 for the electrode holder.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76221-220
- 76221-218
- pH range:
- –2 (min.) – 16 (max.)
- –2 (min.) – 16 (max.)
- mV range:
- –2000 (min.) – 2000 (max.)
- –2000 (min.) – 2000 (max.)
- ISE concentration range:
- 0.01 ppm (ISE) [min.] – 2000 ppm (ISE) [max.]
- 0.01 ppm (ISE) [min.] – 2000 ppm (ISE) [max.]
- Temperature range:
- 32...212 °F (0...100 °C)
- 32...212 °F (0...100 °C)
- Power supply:
- 100 - 240 VAC, 50/60 Hz
- 100 - 240 VAC, 50/60 Hz