VWR® Manual Crimping and Decrimping Tools
Supplier: VWR International
Certificates
About this item
Crimping tools provide secure vial closure
- Easy and convenient handling
- Durable, high quality construction
- Crimper size clearly labeled for quick identification
Screw-stop in crimping handle allows for adjustment of crimping pressure. Decrimping tools provide convenient method for crimp seal removal.