PIPETMAN® Classic™ Single Channel Pipettor, Gilson®
Supplier: GILSON, INC.
Certificates
About this item
Fully adjustable air-displacement pipettor PIPETMAN® Classic™.
- Reliable, accurate, and cost effective
- The original, world-renowned pipetting standard
- PIPETMAN® fixed volume models are also available
The PIPETMAN Classic line, the world standard in pipetting, offers a wide range of continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density. The PIPETMAN Classic line is available in eight single channel models covering a range of volumes from 0.2 µl to 10 ml.
Certifications: Gilson maximum permissible errors are guaranteed only when PIPETMAN® pipettors are used with the recommended PIPETMAN® Diamond Tips.
Customers Who Bought This Also Bought