Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Kemglass Liner, Kewaunee Scientific
Supplier: Avantor
Certificates
About this item
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today. The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
- Un paralleled Venturi enhanced containment technology
- High performance/energy efficient
- Unique ultra-smooth sash system
- Exceptional viewing height
- Flush airfoil
- LED lights with color and intensity adjustability
- Electromechanical sash stops
Color: Light neutral or snow white
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.
Specifications
- Catalog No:
- KV05F282472GNV
- Environmentally Preferable:
- Sash working height:
- 28"
Specifications
Customers Who Bought This Also Bought