Order Entry
ContactUsLinkComponent
Accessories for FastPrep-96™ BigFlex™, MP Biomedicals
Accessories for FastPrep-96™ BigFlex™, MP Biomedicals
Catalog #: CA76045-812
Supplier:  MP Biomedicals
Accessories for FastPrep-96™ BigFlex™, MP Biomedicals
Catalog #: CA76045-812
Supplier:  MP Biomedicals
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BigFlex™ Adapter for FastPrep-96™,8×50 ml
  • Cat. no.:
    CA76045-812
  • Sample Size:
    8 x 50 mL

Specifications

About this item

The BigFlex™ Adapter is a large-scale adapter for the FastPrep-96™ that holds 8 x 50 mL Lysing Matrix tubes.

  • Consistent reproducible results
  • Wide range of disposable 50mL Lysing Matrix tubes available
  • Thorough homogenization of any sample type in seconds

With the FastPrep-96 Instrument, BigFlex adapter and optimized Lysing Matrices, consistent reproducible results are obtained.

BigFlex™ 8 x 50 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at –20°C or –80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

Use with FastPrep-96, MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples at ambient temperatures, for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules and more.

Customers Who Bought This Also Bought