Specifications
- Description:QuickPrep™ All-Metal Sample Holder
- Cat. no.:CA76045-850
Specifications
About this item
All-Metal QuickPrep™ sample holder is the strongest FastPrep adapter used for lysing highly contagious samples and samples suspected of harboring prions.
- High temperature sterilization compatible
- Safest solution for CNS tissue lysis
- High degree of reliability
The superior design and manufacturing of the QuickPrep metal adapter delivers a high degree reliability and ultimate safety facilitating applications involving highly infectious agents.
The QuickPrep™ metal adapter holds 24×2 ml lysing matrix tubes. This is the strongest FastPrep adapter, intended for the most difficult and long-lasting lysis applications. This all-metal adapter can resist high temperatures up to 450 °C and can be sterilized by pyrolysis. Furthermore, the specially designed claws prevent tubes from auto-rotation during lysis, thus allowing continuous tracking of tube labels and/or bar codes. The major use of this adapter is for lysing highly contagious samples and samples suspected of harboring prions. Due to its ability for high temperature sterilization, this adapter is the safest solution for CNS tissue lysis and other applications where prions or other disease causing agents may be present.