PYREX® Transfer Cap, Corning
Catalog #: 10811-374
Supplier: Corning
Specifications
- Description:PYREX® Transfer Cap, Male Luer Lock, 5 L
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:10811-374
- Supplier No.:11503
Specifications
About this item
Transfer Cap for optimum growth with with 1/4" Barb Connection.
- Sterile
Customers Who Bought This Also Bought