Specifications
- Description:Spherical Glass Plating Beads
Specifications
About this item
Roll & Grow™ Plating Beads are used to spread 25–400μL of bacterial or yeast cells across the surface of an agar plate.
- Sterile
- Easy to use
- Spread cells in just seconds per plate
- Provides even colony distribution
- Eliminates hazards of flame and ethanol
Roll & Grow™ Plating Beads provide a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow® Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.