ContactUsLinkComponent
Memmert CO₂ Incubator ICO Series
Memmert CO₂ Incubator ICO Series
Supplier:  Memmert
   Sustainable Options Available
Certificates
Memmert CO₂ Incubator ICO Series
Memmert CO₂ Incubator ICO Series
Supplier:  Memmert
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 76183-542
  • 76183-540
  • 76183-538
  • 76183-536
  • Model:
  • ICO240
  • ICO150
  • ICO105
  • ICO50
  • Environmentally Preferable:
  • Sustainable Packaging, Low Manufacturing Impact
  • Sustainable Packaging, Low Manufacturing Impact
  • Sustainable Packaging, Low Manufacturing Impact
  • Sustainable Packaging, Low Manufacturing Impact
  • Recovery time after 30 second door opening at 37 °C:
  • 6 min
  • 5 min
  • 4 min
  • 3 min
  • Usable volume:
  • 118.85 L
  • 69.89 L
  • 43.01 L
  • 19.67 L
  • CO₂ recovery time after 30 second door opening at 5 Vol.% CO₂:
  • 9 min
  • 8 min
  • 6 min
  • -
  • Energy consumption:
  • 85 Wh/h at 37 °C
  • 80 Wh/h at 37 °C
  • 60 Wh/h at 37 °C
  • 60 Wh/h at 37 °C
  • Environmental certification:
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)