Axygen® Automation Tips for PerkinElmer® Janus®/MultiProbe, Corning
Supplier: Corning
Certificates
About this item
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
- Engineered to the exact specifications
- Complete protection against contamination
Axygen® offers a comprehensive line of tips for PerkinElmer® automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are certified RNase-/DNase-free and nonpyrogenic. Available in nonsterile and sterile formats. Clear and Conductive Black polypropylene tips are available in 96-well format designed for PerkinElmer® Janus®/MultiProbe Variospan-1, 4, 8 heads.
Certifications: Tips are certified RNase-/DNase-free, and nonpyrogenic.
Specifications
- Catalog No:
- 89080-542
- 89080-540
- 89080-538
- 89080-536
- 89080-554
- 89080-552
- 89080-550
- 89124-954
- 89080-546
- 89080-544
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa