Microscope Slide Mailer for 5 Slides with Side Opening, Globe Scientific
Catalog #: 75845-146
Supplier: Globe Scientific
Specifications
- Description:Polypropylene Slide Mailer with Side Opening
- Cat. no.:75845-146
- Supplier No.:513065
Specifications
About this item
The side opening lid provides easy access to the entire length of the slide (not just the end)
Slide mailers are produced from polypropylene and feature inner slots which keep slides separated and secure during transport.
Customers Who Bought This Also Bought