ContactUsLinkComponent
VWR® Septa Sealing Mat
VWR® Septa Sealing Mat
Catalog #: 470166-590
Supplier:  VWR International
VWR* Sapphire Advance 96-well Sealing Mat
VWR® Septa Sealing Mat
Catalog #: 470166-590
Supplier:  VWR International
Specifications
  • Description:
    Septa Sealing Mat
Specifications
About this item
This 96-well septa sealing mat is a cost-effective alternative and is compatible with the ABI 3100.
  • 96-well sealing mat with septa
  • Round well
  • Non-perforated
The septa sealing mat is compatible for use on the ABI 3100. Standard packaging includes 10 mats/pack, 5 packs/case. In addition to the standard packaging option, it is also available in packs of 10.
Customers Who Bought This Also Bought