Digi-Sense® Benchtop Ramp/Soak Controllers, Cole-Parmer
Supplier: Argos Technologies
Certificates
About this item
The ideal controller for processes requiring several steps.
- Models available for type J, K, or T thermocouples, or RTD probes
- Software allows for data point collection and custom designed interface
- Program up to a 40-step profile
Ordering information: Includes software, 6 ft. detachable power cord, and a lattice support bracket.
Specifications
- Catalog No:
- 76183-326
- 76183-324
- 76183-630
- 76183-628
- Temperature range:
- Depends on temperature probe (order separately)
- Depends on temperature probe (order separately)
- Depends on temperature probe (order separately)
- Depends on temperature probe (order separately)
- Temperature accuracy:
- 0.1% of span, ±1.0 °F/°C
- 0.1% of span, ±1.0 °F/°C
- 0.1% of span, ±1.0 °F/°C
- 0.1% of span, ±1.0 °F/°C
- Control type:
- Output 1: Solid State Relay, PID with Ramp and Soak
- Output 1: Solid State Relay, PID with Ramp and Soak
- Output 1: Solid State Relay, PID with Ramp and Soak
- Output 1: Solid State Relay, PID with Ramp and Soak
- Display type:
- LED
- LED
- LED
- LED
Specifications
Customers Who Bought This Also Bought