Specifications
- Description:Stain Kit
- Size:3 bottles per kit
- Cat. no.:CA10842-586
- Supplier No.:ES3405
Specifications
About this item
AFB stain kit for the histological visualization of acid fast organisms.
- For in-vitrodiagnostic use only
- 15-minute processing time
Intended for use in the histological visualization of Acid Fast Bacteria and Tubercle Bacilli. The Acid Fast Bacteria (AFB) stain kit includes all the required solutions to perform the Acid Fast staining procedure including a carbol fuchsin solution, an acid alcohol solution (0.5%) and a light green solution.