Specifications
- Description:Micro 4 High-Content Imaging System
- Cat. no.:CA76177-142
Specifications
About this item
Position your research with breakthrough, intelligent imaging solutions. The ImageXpress® Micro 4 High-Content Imaging System offers an agile solution enabling you to expedite your research.
- Faster-than-ever, agile system, which grows with your needs
- Acquire statistically relevant data faster with large field of view and >3 log dynamic range
- Expand your research capabilities with available options
- High-content acquisition and image analysis software for complex biological assays
4th Generation Platform with unlimited configurability, developed to address your research needs.
Minimize tiling effect and reduce your acquisition time with the ability to image a whole 384 well at once.
MetaXpress® Software is optimized to perform with the ImageXpress Micro imaging systems, giving you precise control over image acquisition and powerful, elegant tools for analysis. Gain speed and convenience with our single wizard to guide you through image acquisition and analysis, laser-based autofocus, pre-designed image analysis application modules, parallel processor support, and automatic data transfer scheduling. An interactive custom module editor transforms your knowledge, ideas, and intuition into entirely new, multiple-step analysis routines that can be created and run in minutes.
The ImageXpress® Micro 4 High-Content Imaging System represents the culmination of four generations of imaging expertise. The latest agile design allows you to boost your research with this faster-than-ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
Built on over 30 years of cell-based imaging experience, the ImageXpress Micro 4 High-Content Imaging system is engineered for performance to reliably accelerate the pace of your research.