SP Wilmad-LabGlass Reaction Monitoring System NMR Tube, 5 mm, SP Industries
Supplier: SP Industries
Certificates
About this item
Designed to monitor reactions from start to finish, the Wilmad-LabGlass Reaction Monitoring System is a two-chamber borosilicate glass NMR tube that allows for in-tube mixing.
- Allows for acquisition of both pre-mix and post-mix spectra
- In-situ mixing of solutions provides a clear ‘before and after’ reaction picture
- Enables specific reaction endpoint determination
- Eliminates variables and reduces risk of contamination during experiments
- Permits researchers to run reaction experiments without specialized instruments or equipment
- Made from Type 1, Class A Borosilicate Glass
The system features an inner chamber with a Teflon® tip that when secured creates a positive seal separating solutions until the user is ready to mix.
The black phenolic cap has a white rubber liner for the outer tube allowing for easy sample storage.
Tubes may allow for capture of initial kinetic data points in benchtop spectrometers where the upper portion of the sample tube is accessible.
Delivery: Includes an extra black phenolic cap.
Caution: Not for use in spinning experiments. The black phenolic cap used during mixing does not have a liner and should always be kept in a vertical position.
Customers Who Bought This Also Bought