Azlon® Economy Molded Cylinders, Polymethylpentene, Dynalon
Supplier: Dynalon
Certificates
About this item
Clear, shatterproof polymethylpentene polymer cylinder.
- Glass-like clarity of polymethylpentene (PMP) construction
- Shatter-resistant design reduce breakage and incidence of injury
- Chemically resistant to most acids, bases, and many common solvents
- Autoclavable
Plastic cylinders feature permanently molded graduations that will not fade after repeated autoclaving. Meets stringent international quality criteria according to ISO 6706 standards. Tolerances according to DIN 12 680 TT, Class B. Maximum temperature is 135 °C.
Certifications: Meets ISO 6706 standards
Customers Who Bought This Also Bought