To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG2b Isotype Control (LTF-2)
- Clonality:Monoclonal
- Clone:LTF-2
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Cat. no.:75842-446
- Supplier No.:41823-25-100
Specifications
About this item
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse