To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mouse
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Cat. no.:75841-352
- Supplier No.:44212-80-100
Specifications
About this item
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse