To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:2A3
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Conjugation:PE (Phycoerythrin)-Cy5®
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Cat. no.:75842-434
- Supplier No.:41813-65-100
Specifications
About this item
The 2A3 antibody was obtained by hybridization of MouseNS-1 myeloma cells with spleen cells harvested from BALB/c mice immunized with phytohemagglutinin-activated human T cells. The 2A3 antibody is a CD25 clone and used as an Isotype Control for rat IgG2a antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: 2A3
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse