Order Entry
ContactUsLinkComponent
Anti-TNFA Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MP6-XT22]
Anti-TNFA Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MP6-XT22]
Catalog #: 75841-394
Anti-TNFA Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MP6-XT22]
Catalog #: 75841-394
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor alpha
  • Clonality:
    Monoclonal
  • Clone:
    MP6-XT22
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Antigen symbol:
    TNFA
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Size:
    25 μg
  • Form:
    liquid
  • Antigen synonyms:
    Tumor Necrosis Factor alpha|DIF|Differentiation inducing factor|MCF|Necrosin|TNFSF2|Cachectin|Macrophage cytotoxic factor
  • Cat. no.:
    75841-394
  • Supplier No.:
    84422-77-25

Specifications

About this item

The MP6-XT22 monoclonal antibody specifically reacts with mouse tumor necrosis factor alpha (TNF-alpha), a 156 amino pro-inflammatory cytokine. It is secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. NF-α is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. The MP6-XT22 is reported to neutralize the bioactivity of natural or recombinant TNF-alpha and is useful for intracellular immunofluorescent staining.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse

Customers Who Bought This Also Bought