To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Toll-like receptor 9
- Clonality:Monoclonal
- Clone:M9.D6
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Antigen symbol:TLR9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:toll-like receptor 9|TLR-9
- Cat. no.:75843-288
- Supplier No.:31912-50-25
Specifications
About this item
The M9.D6 monoclonal antibody specifically reacts with mouse CD289 also known as toll-like receptor 9 (TLR9). CD289 is involved in the activation of innate immunity, in acquired immune responses, and autoimmune diseases. It is involved in the immune system’s response to unmethylated CpG dinucleotide sequences such as those found in bacterial, viral, or synthetic DNA. CD289 assists in pathogen recognition and is expressed by subtypes of dendritic cells and B cells.
Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone: M9.D6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse