To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Toll-like receptor 4
- Clonality:Monoclonal
- Clone:MTS510
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:TLR4
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:TLR4/ MD-2|toll-like receptor MD2|TLR-4/ MD2
- Cat. no.:75842-524
- Supplier No.:74812-25-100
Specifications
About this item
The MTS510 monoclonal antibody specifically reacts with the Toll-Like Receptor 4 (TLR-4) and MD-2 (LY96) molecular complex. The complex of TLR-4, MD-2, and CD14 regulates the innate immune system recognition of bacterial lipopolysaccharides (LPS) and is expressed on the surface of thioglycollate-elicited macrophages. The epitope that binds the MTS510 antibody is lost after LPS stimulation and the antibody can be used to co-immunoprecipitates MD-2 and TLR4.
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: MTS510
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse