To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:TER-119
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:TER-119
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Ly-76|TER119|Ly76|Erythroid cell marker
- Cat. no.:75843-546
- Supplier No.:81212-50-100
Specifications
About this item
The TER-119 monoclonal antibody specifically binds to the mouse 52 kDa Glycophorin A expressed on the erythroid lineage cells, from the early proerythroblast to mature erythrocyte stages, in the fetal liver, adult bone marrow, newborn liver, yolk sac, and adult lymphoid organs. The TER-119 antibody is used, together with other antibodies, as a marker for hematopoietic lineages cells.
Type: Primary
Antigen: TER-119
Clonality: Monoclonal
Clone: TER-119
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse