To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD279 (PD-1)
- Clonality:Monoclonal
- Clone:MIH4
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:PD-1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:B7h.5|VTCN1
- Cat. no.:75844-498
- Supplier No.:31811-50-25
Specifications
About this item
The MIH4 monoclonal antibody specifically reacts with human Programmed death-1 (PD-1 or CD279), a 50-55 kDA glycoprotein. It is expressed on mainly on activated B, T, and myeloid cells. Within the cytoplasmic region, PD-1 contains an Immunoreceptor tyrosine-based inhibitory motif (ITIM) and seems to regulate peripheral tolerance. The lack or mutation of CD279 is linked to autoimmune disorders.
Type: Primary
Antigen: PD-1
Clonality: Monoclonal
Clone: MIH4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human