To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:OCT3/4
- Clonality:Monoclonal
- Clone:EM92
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:OCT3/4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:octamer-binding transcription factor|OCT4|OCT3|POU5F1
- Cat. no.:75844-512
- Supplier No.:78011-60-25
Specifications
About this item
The EM92 monoclonal antibody reacts with human and mouse Oct 3/4, a POU family transcription factor. Oct 3/4 is encoded by the Pou5F1 gene and expressed on undifferentiated cells and lost in adult tissue. Fbx15, FGF-4, NANOG, Rex1, and UTF1 expression is regulated with Oct 3/4 and in many cases gene expression is regulated in concert with Sox2.
Type: Primary
Antigen: OCT3/4
Clonality: Monoclonal
Clone: EM92
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse