To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Killer cell lectin-like receptor subfamily B, member 1
- Clonality:Monoclonal
- Clone:PK136
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:KLRB1
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:NKR-P1C|NKR-P1B|CD161c|Ly-55|CD161b|CD161
- Cat. no.:75842-642
- Supplier No.:83712-80-100
Specifications
About this item
The PK136 monoclonal antibody is specific for the mouse NK1.1, a receptor from the killer cell lectin-like receptor (KLR) family. Nk1.1 is an antigen encoded by the Klrb1c/NKR-P1C gene expressed by the natural killer cells of some selected strains of mice (C57BL, FVB/N, NZB) and encoded by the Klrb1b/NKR-P1B gene expressed on Swiss NIH and SJL mice. Pk136 binds to an epitope common to NKR-P1B and NKR-P1C.The Klrb1 is a family of type II transmembrane C-type lectin receptors. Klrb1c activates the NK-cell cytotoxicity, while Klrb1b inhibits it. PK136 is useful in defining the NK cells and the rare population of NK-T lymphocytes and specific cultured monocytes.
Type: Primary
Antigen: KLRB1
Clonality: Monoclonal
Clone: PK136
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse