To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:MHC Class I
- Clonality:Monoclonal
- Clone:34-1-2S
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:MHCI
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:H2Kd|MHCI|H2Dd
- Cat. no.:75841-424
- Supplier No.:86112-50-100
Specifications
About this item
The 34-1-2S monoclonal antibody specifically reacts with the mouse H-2k/H2D MHC class I alloantigens, which are involved in antigen presentation to T cells. The 34-1-2S antibody cross-reacts with the b, p, q, r, and s haplotypes.
Type: Primary
Antigen: MHCI
Clonality: Monoclonal
Clone: 34-1-2S
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse