To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:1A8
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:Ly-6G
- Flow cytometry:Yes
- Size:100 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Ly6G
- Cat. no.:75843-616
- Supplier No.:83112-25-100
Specifications
About this item
The 1A8 monoclonal antibody specifically reacts with the 21-25 kDa glycophosphatidylinositol-anchored protein known as Ly-6G, expressed by the granulocytes from the bone marrow and periphery neutrophils. Ly-6G and Ly-6C form the Granulocyte Receptor-1 antigen (GR-1). The binding of the 1A8 antibody to the Ly-6G can be blocked by another antibody, RB6-8C5, which also recognizes Ly-6C. While 1A8 is specific for only Ly-6G, RB6-8C5 also binds to Ly-6C.
Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse