To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:D7
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:Ly-6A/E
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Size:50 μg
- Form:liquid
- Antigen synonyms:Ly6A|Ly6E|Sca1|Ly6AE
- Cat. no.:75843-610
- Supplier No.:82912-77-50
Specifications
About this item
The D7 monoclonal antibody specifically reacts with mouse Ly-6A/E (Sca-1), a 18 kDa GPI-linked surface protein. It is expressed on multipotent hematopoietic stem cells (HSC) and subsets of B and T cells and involved in B and T cell lymphocyte responses. Splenocytes from Ly-6.2 strains exhibit a larger number of D7-positive cells than those from the Ly-6.1 strains.
Type: Primary
Antigen: Ly-6A/E
Clonality: Monoclonal
Clone: D7
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse