To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin beta 1
- Clonality:Monoclonal
- Clone:TS2/16
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:ITGB1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Size:25 tests
- Form:liquid
- Antigen synonyms:Fibronectin Receptor Beta|CD29
- Cat. no.:75842-160
- Supplier No.:11311-50-25
Specifications
About this item
The TS2/16 moncolonal antibody specifically reacts with human CD29 (Integrin beta1), a 130 kDA type I glycoprotein expressed on hematopoietic and non-hematopoietic cells. It forms the VLA-(1-6) molecules with integrin alpha-(1-6) and is involved in the adhesion between cell-cell and cell-matrix. The TS2/16 antibody has been found to activate beta 1 integrins.
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: TS2/16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human