To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD127 (IL-7Ra) (A7R34)
- Clonality:Monoclonal
- Clone:A7R34
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:IL-7Ra
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Size:100 μg
- Form:liquid
- Antigen synonyms:Interleukin-7 Receptor alpha|IL-7Ra
- Cat. no.:75844-484
- Supplier No.:19212-80-100
Specifications
About this item
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the α subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127. 
Type: Primary
Antigen: IL-7Ra
Clonality: Monoclonal
Clone: A7R34
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse