Order Entry
ContactUsLinkComponent
Anti-IL-4 (11B11) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11B11]
Anti-IL-4 (11B11) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11B11]
Catalog #: 75843-516
Anti-IL-4 (11B11) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11B11]
Catalog #: 75843-516
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL-4 (11B11)
  • Clonality:
    Monoclonal
  • Clone:
    11B11
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse
  • Antigen symbol:
    IL-4 (11B11)
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Size:
    25 μg
  • Form:
    liquid
  • Antigen synonyms:
    IL4|Macrophage fusion factor (MFF)|T cell growth factor-2 (TCGF-2)|BSF-1|Interleukin-4|Mast cell growth factor-2 (MCGF-2)
  • Cat. no.:
    75843-516
  • Supplier No.:
    81112-60-25

Specifications

About this item

The 11B11 monoclonal antibody specifically reacts with mouse 14 kDa cytokine IL-4, expressed by the activated NKT cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen.The 11B11 antibody neutralizes the IL-4 cytokine.

Type: Primary
Antigen: IL-4 (11B11)
Clonality: Monoclonal
Clone: 11B11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse

Customers Who Bought This Also Bought