Order Entry
ContactUsLinkComponent
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]
Catalog #: 75843-524
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2]
Catalog #: 75843-524
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL-4
  • Clonality:
    Monoclonal
  • Clone:
    MP4-25D2
  • Host:
    Rat
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Antigen symbol:
    IL-4
  • Flow cytometry:
    Yes
  • Size:
    500ug
  • Format:
    SAFIRE Purified
  • Form:
    liquid
  • Antigen synonyms:
    IL4|B-cell stimulating factor-1|Interleukin-4
  • Cat. no.:
    75843-524
  • Supplier No.:
    81121-25-500

Specifications

About this item

The MP4-25D2 monoclonal antibody specifically reacts with human Interleukin-4 (IL-4), expressed by the activated T cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen. The MP4-25D2 is a neutralizing antibody and is reported to be cross-reactive with rhesus monkeys.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Human

Customers Who Bought This Also Bought