To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-4 (11B11)
- Clonality:Monoclonal
- Clone:11B11
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:IL-4
- Flow cytometry:Yes
- Size:500 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:IL4|Macrophage fusion factor (MFF)|T cell growth factor-2 (TCGF-2)|BSF-1|Interleukin-4|Mast cell growth factor-2 (MCGF-2)
- Cat. no.:75843-512
- Supplier No.:81112-25-500
Specifications
About this item
The 11B11 monoclonal antibody specifically reacts with mouse 14 kDa cytokine IL-4, expressed by the activated NKT cells and mast cells. IL-4 is species-specific and stimulates the proliferation and differentiation of B lymphocytes. It upregulates the expression of IgE receptors and class II MHC antigen.The 11B11 antibody neutralizes the IL-4 cytokine.
Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: 11B11
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse