To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-2 (JES6-5H4)
- Clonality:Monoclonal
- Clone:JES6-5H4
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Antigen symbol:IL-2
- Conjugation:Biotin
- Flow cytometry:Yes
- Size:25 μg
- Form:liquid
- Antigen synonyms:Interleukin-2|Eosinophil differentiation factor (EDF)|Macrophage-activating factor for cytotoxicity I (MAF-C I)|Thymocyte differentiation factor (TDF)|Killer cell helper factor (KHF)|T cell growth factor (TCGF)
- Cat. no.:75841-458
- Supplier No.:86412-30-25
Specifications
About this item
The JES6-5H4 antibody specifically binds to mouse IL-2, a 17 kDa immunoregulatory cytokine and growth factor for T and B lymphocytes, NK cells, monocytes, and macrophages. IL-2 induces the cytotoxic operations of NK cells, activates B cells, and plays a role in the differentiation of T cells. JES6-5H4 seems to have a neutralizing effect on interleukin-2.
Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse